L(+)-Asparagine monohydrate
Supplier: Serva
Certificates
About this item
CAS Number:5794-13-8
:
Formula:C₄H₈N₂O₃·1H₂O
MDL Number:MFCD00151038
:150.13 g/mol
Merck Index:14,00837
Melting Pt:215…217 °C
Density:1.567 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-735-9