Order Entry
Germany
ContactUsLinkComponent
L(+)-Asparagine monohydrate
L(+)-Asparagine monohydrate
Catalog # MOLEM86033702
Supplier:  Molekula
CAS Number:  
L(+)-Asparagine monohydrate
Catalog # MOLEM86033702
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :150.13 g/mol
Merck Index:14,00837
Melting Pt:215…217 °C
Density:1.567 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-735-9
CAS Number:5794-13-8
Formula:C₄H₈N₂O₃·1H₂O
MDL Number:MFCD00151038