L(+)-Asparagine monohydrate
Catalog # BACE4030374.0250
Supplier: BACHEM BIOCHEMICA
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:150.13 g/mol
Merck Index:14,00837
Melting Pt:215…217 °C
Density:1.567 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-735-9
CAS Number:5794-13-8
Formula:C₄H₈N₂O₃·1H₂O
MDL Number:MFCD00151038