Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Paraformaldehyde
Paraformaldehyde, Sigma-Aldrich®
Paraformaldehyde, Sigma-Aldrich®
:
Merck
Paraformaldehyde, Sigma-Aldrich®
:
Merck
:
30525-89-4
:
:
(CH₂O)n
:
MFCD00133991
:
13,07096
:
135…136 °C (13 mmHg)
:
123 °C
:
1.46 g/cm³ (20 °C)
:
70 °C
:
Ambient
:
4.1,III
:
M-Clarity™ quality level = MQ200
:
2213
:
GHS02
:
GHS08
:
GHS07