


Paraformaldehyde, Sigma-Aldrich®
Catalog # SIAL16005-6X1KG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:6 kg
- Seq:4122
Specifications
About this item
:
Merck Index:13,07096
Boiling Pt:135…136 °C (13 mmHg)
Melting Pt:123 °C
Density:1.46 g/cm³ (20 °C)
Flash Pt:70 °C
Storage Temperature:Ambient
UN:2213
ADR:4.1,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS08
:GHS07
CAS Number:30525-89-4
Formula:(CH₂O)n
MDL Number:MFCD00133991