Order Entry
Germany
ContactUsLinkComponent
 
Irritant
Corrosive
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Proteomics Grade
Catalog # M144-1L
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Proteomics Grade
Catalog # M144-1L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Cloud Point (1%, Water):
    22 - 28 °C
  • Colour (APHA):
    < 100
  • Refractive index:
    1,48 - 1,50

Specification Test Results

About this item

 :
Boiling Pt:150 °C (1013hPa)
Density:1,055 g/cm³ (20 °C)
Flash Pt:93 °C (closed cup)
ADR:9,III
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:9036-19-5
MDL Number:MFCD00128254