TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9002-93-1
:9036-19-5
MDL Number:MFCD00128254
Boiling Pt:∼150 °C (1013 hPa)
Density:1.055 g/cm³
Flash Pt:254 °C
ADR:9,III
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear to slightly hazy liquid
- Infrared spectrum:Conforms
- Refractive index:1.4880 to 1.4950 (20°C, 589 nm)
- Color scale:≤100 APHA
- Cloud point:22 to 28°C (1 % in water)