Order Entry
Germany
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Millipore®
Corrosive
Irritant
Environment
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Millipore®
Catalog # 648463-50
CAS Number:  
Corrosive
Irritant
Environment
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Millipore®
Catalog # 648463-50
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
Merck Index:13,06793
Boiling Pt:∼200 °C (1013 hPa)
Melting Pt:6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:251 °C (closed cup)
UN:3082
ADR:9,III
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Refrigerator
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505