Polydimethylsiloxane, 10 cSt DC 200 Fluid
Catalog # XXXXDC200.9018
Supplier: Chemicals
CAS Number:
Specifications
- Pk:18 kg
- Seq:4235
Specifications
About this item
:
CAS Number:63148-62-9
:9016-00-6
Formula:(CH₃)₃SiO(Si(CH₃)₂O)n
MDL Number:MFCD00084411