Order Entry
Germany
ContactUsLinkComponent
Aluminium standard solution, 1000 mg/L Al in dil. nitric acid (from Al) standard for AAS and ICP
Aluminium standard solution, 1000 mg/L Al in dil. nitric acid (from Al) standard for AAS and ICP
Catalog # JOBIPLAL2-2T
CAS Number:  
Aluminium standard solution, 1000 mg/L Al in dil. nitric acid (from Al) standard for AAS and ICP
Catalog # JOBIPLAL2-2T
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2-3% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item

Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.