Cleaning spray for electronics, KONTAKT WL
Catalog # 291-1246
Supplier: HM INDUSTRIESERVICE
Specifications
- Description:Cleaner spray for electronic parts, 200 ml
- Pk:200 mL
Specifications
About this item
KONTAKT WL is the universal spray wash for all electronics. The solvent mixture removes a wide range of dirt such as oil, grease, soot or nicotine residues from electrical devices, plugs, engines or precision mechanical gears.
KONTAKT WL has a high dissolving power, good material compatibility with common electronic components and dries quickly and without leaving any residue.