Order Entry
Germany
ContactUsLinkComponent
Bovine serum albumin (BSA), Fraction V
Bovine serum albumin (BSA), Fraction V
Supplier:  Merck
Certificates
Bovine serum albumin (BSA), Fraction V
Supplier:  Merck

Specifications

  • CAS No.:
    9048-46-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Purity:
    >98%
  • pH:
    pH 7

Specifications