ContactUsLinkComponent
Detergents for glasswares, NOCHROMIX®, Sigma-Aldrich®
Detergents for glasswares, NOCHROMIX®, Sigma-Aldrich®
Catalog # SIAL328693-10PAK
Supplier:  Merck
Detergents for glasswares, NOCHROMIX®, Sigma-Aldrich®
Catalog # SIAL328693-10PAK
Supplier:  Merck
Specifications
  • Description:
    NOCHROMIX® cleaning agent
  • Pk:
    10
Specifications
About this item
A metal-free white powder when mixed with sulfuric acid, makes 2,3 l of cleaning solution for laboratory glasswares.
  • Non-polluting
  • Non-carcinogenic
Certifications: EPA recommended.