0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Proteins and Peptides
Human albumin (from serum)
Human albumin (from serum)
:
Merck Millipore (Calbiochem)
Human albumin (from serum)
:
Merck Millipore (Calbiochem)
CAS No.:
70024-90-7
Source:
Serum
Species:
Human
Storage conditions:
Refrigerator
Protein synonyms:
Human Serum Albumin|HSA|serum albumin|HAS|serum of albumin
Protein/peptide name:
Albumin