Order Entry
Germany
ContactUsLinkComponent
Pig Gelatine, Type A (from Skin), MP Biomedicals
Pig Gelatine, Type A (from Skin), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Pig Gelatine, Type A (from Skin), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9000-70-8
  • Source:
    Skin
  • Species:
    Pig
  • Storage conditions:
    +15 to +30 °C
  • Form:
    Powder
  • Protein synonyms:
    Type A|Gelatin
  • Protein/peptide name:
    Gelatine, Type A
  • Colour:
    Yellow
  • pH:
    4,5 - 5,9
  • Grade:
    Food grade

Specifications