Glucose oxidase (from Aspergillus niger)
Catalog # 345386-10
Supplier: Merck Millipore (Oncogene)
CAS Number:
Specifications
- Pk:10 KU
- CAS No.:9001-37-0
- Source:Aspergillus niger
- Storage conditions:Freezer
- MDL No.:MFCD00131182
- Enzyme name:Glucose oxidase