

3-Mercapto-1,2-propanediol ≥90% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-27-5
:
Formula:C₃H₈O₂S
MDL Number:MFCD00004879
:108.16 g/mol
Merck Index:14,9335
Boiling Pt:117…119 °C (5 mmHg)
Density:1.247
Flash Pt:149 °C (300 °F)
EINECS:202-495-0
:GHS06
:GHS07
Storage Temperature:Ambient