DL-Sodium lactate 50% in aqueous solution Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:72-17-3
:312-85-6
Formula:H₃CCH(OH)COONa
MDL Number:MFCD00065400
:112.06 g/mol
Density:1.263 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-772-0
Specification Test Results
- Assay (Sodium 2-hydroxypropanoate):Min. 50%
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification B:Passes test
- Appearance of solution:Passes test
- pH:6.5 - 9.0
- Reducing sugars and sucrose:Passes test
- Chlorides (calc. ref. to Na lactate):Max. 50 ppm
- Oxalates and phosphates:Passes test
- Sulphates (calc. ref. to Na lactate):Max. 100 ppm
- Iron (calc. ref. to Na lactate):Max. 10 ppm
- Heavy metals (calc. ref. to Na lactate):Max. 10 ppm
- Residual solvents:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000)
Packaging Information
27927_SODIUM LACTATE 50% PH.EUR_BSE_REV0
BSE/TSE Certificates
27927_SODIUM LACTATE 50% PH.EUR_LATEX_REV0
Additional Product Details
spec_VWRC27927.298 spec_VWRC27927.367
Product specification
VWR Chemicals for Biopharma and Pharma Production