L(+)-Sodium lactate solution ∼50%, EMPROVE® EXPERT Ph. Eur., BP, USP suitable for use as excipient, SAFC®
: Merck
:867-56-1
:
:C₃H₅NaO₃
:MFCD00066576
:112.06 g/mol
:109 °C (1013 hPa)
:1.263 g/cm3 (20 °C)
:M-Clarity™ quality level = MQ500 (more info.)
:Ambient
:200-772-0
- Assay (Perchloric acid titration)#:50.0 - 51.0 %
- Assay (Stereochemical purity of Sodium-(S)-lactate)#:≥ 95.0 %
- Identity (Sodium):passes test
- Identity (Lactat):passes test
- Appearance of solution:Clear and not more intense in color than reference solution BY₇
- pH (in present substance):6.5 - 9.0
- Chloride (Cl)#:≤ 50 ppm
- Sulfate (SO₄) (Ph Eur)#:≤ 100 ppm
- Sulfate (SO₄) (USP):passes test
- Al (Aluminium):≤ 0.1 ppm
- As (Arsenic)#:≤ 3 ppm
- Fe (Iron)#:≤ 10 ppm
- Hg (Mercury):≤ 1 ppm
- Pb (Lead):≤ 5 ppm
- Oxalate and Phosphate (Ph Eur):passes test
- Citrate, Oxalate, Phosphate or Tartrate (USP):passes test
- Methanol and Methyl Esters:≤ 250 ppm
- Reducing sugars and sucrose (Ph Eur):passes test
- Sugar (USP):passes test
- Methanol#:≤ 50 ppm
- Residual solvents (ICH Q3C):excluded by manufacturing process
- Total aerobic microbial count (TAMC):≤ 100 CFU/ml
- Total combined yeasts/moulds count (TYMC):≤ 10 CFU/ml
- Bile-tolerant gram-negative bacteria (absent in 1 ml):passes test
- Candida albicans (absent in 1 ml):passes test
- Escherichia coli (absent in 1 ml):passes test
- Pseudomonas aeruginosa (absent in 1 ml):passes test
- Salmonella (absent in 10 ml):passes test
- Staphylococcus aureus (absent in 1 ml):passes test
- Endotoxins:< 5