L(+)-Sodium lactate 60% (w/w) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:867-56-1
:
Formula:C₃H₅NaO₃
MDL Number:MFCD00066576
:112.06 g/mol
Density:1.33 g/cm³
Storage Temperature:Ambient
EINECS:212-762-3
Specification Test Results
- Appearance:Clear viscous liquid
- Refractive index:1.4220 to 1.4250 (20°C, 589 nm)
- Density:(20°C) 1.32 to 1.34 g/ml
- Color scale:≤50 APHA
- pH:6.5 to 8.5
- Titration with HCl:59 to 61 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤20 ppm
- Iron (Fe):≤10 ppm
- Sodium (Na):12.1 to 12.6 %
- Heavy metals (as Pb):≤10 ppm
- Isomer:≥97 % L(+) (Stereochemical purity)