Order Entry
Germany
ContactUsLinkComponent
Agarose
Agarose
Catalog # VECTSA-5010-2
CAS Number:  
undefined
Agarose
Catalog # VECTSA-5010-2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
Melting Pt:60…90 °C
Storage Temperature:Ambient
EINECS:232-731-8
CAS Number:9012-36-6
MDL Number:MFCD00081294