Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Agarose

Marketsource
Supplier:   TREVIGEN
Agarose
Supplier:  TREVIGEN
Marketsource
Certificates
Agarose

4 Options Available Below

Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource