Order Entry
Germany
ContactUsLinkComponent
Needles, STERITEX®
Needles, STERITEX®
Catalog # CODA646008
Supplier:  CODAN
undefined
Needles, STERITEX®
Catalog # CODA646008
Supplier:  CODAN

Specifications

  • Gauge:
    18
  • Description:
    Air inlet needle with non wetable filter, STERITEX®
  • Length:
    80 mm
  • Pk:
    50

Specifications

About this item

This needle is only used for the inlet of air into bottles.

  • Metal needle with non return valve
  • With 0,2 µm air filter
Caution: Do not use for reconstitution of lyophilisate.