Orders LinkContactUsLinkComponent
Transport boxes, Securipack
 :   E3 Cortex
Transport boxes, Securipack
 :  E3 Cortex
Transport boxes, Securipack
Transport boxes, Securipack
This Securipack packages are ready for use in the transport of samples of hazardous materials, except for Classes 7 and 6.2.
  • Very simple to use, this package eliminates risk posed by error or oversight
  • Mechanical high strength, in particular in the stacking by the juxtaposition of double walls on all the faces
  • High absorption power of the shocks on all the faces, guaranteeing a perfect protection in case of an accidental fall
Universal application for products regulated except for the radioactive and infectious products.
Protects against the following risks: It is a good oxidiser; non flammable and non toxic; corrosive
 : Supplied with 1 case (sheath), 2 crossed envelopes dressed in a foam thickness, 1 pocket of metallic or plastic retention and 1 sluggish absorbing product. Interior packaging and hazard class label may be supplied upon request.
Product Family Options
  • Capacity
    Description
    W×D×H ext.
    W×D×H int.
  • 1 L
    Transport box for samples, load capacity 5,0 kg
    220×220×360 mm
    140×140×230 mm
    1 * 1 ST
    1 * 1 ST - 98,20 €
    98,20 €
    1 * 1 ST
       98,20 €
  • 0,3 L
    Transport box for samples, load capacity 0,92 kg
    130×130×215 mm
    80×80×140 mm
    1 * 1 ST
    1 * 1 ST - 59,10 €
    59,10 €
    1 * 1 ST
       59,10 €
  • 0,5 L
    Transport box for samples, load capacity 2,7 kg
    180×180×315 mm
    130×130×220 mm
    1 * 1 ST
    1 * 1 ST - 77,90 €
    77,90 €
    1 * 1 ST
       77,90 €
  • 2,5 L
    Transport box for samples, load capacity 12,3 kg
    255×255×450 mm
    180×180×330 mm
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • 5 L
    Transport box for samples, load capacity 13,7 kg
    310×310×430 mm
    230×230×300 mm
    1 * 1 ST
    1 * 1 ST - 183,00 €
    183,00 €
    1 * 1 ST
       183,00 €