ContactUsLinkComponent
Cleanroom wipes, Clino® Carbon
Cleanroom wipes, Clino® Carbon
Catalog # PPSA3600098
undefined
Cleanroom wipes, Clino® Carbon
Catalog # PPSA3600098
Specifications
  • Packed:
    5/pack
  • Colour:
    White
  • L×W:
    350×340 mm
  • Pk:
    5
Specifications
About this item
Reusable polyester/carbon cloth for use on all smooth and water-resistant surfaces.
  • Low particle emission
  • Electrostatically conductive
  • Autoclavable
Suitable for use in ISO class 5 - 8 (GMP grade A/B, C, D) cleanrooms.