Agarose 15, Electran for electrophoresis
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:9012-36-6
:
MDL Number:MFCD00081294
Melting Pt:60…90 °C
Storage Temperature:Ambient
EINECS:232-731-8
Specification Test Results
- Gel strength (1.5 %; water):Min. 1800 g/cm²
- Gelling temperature (1,5 %; water):Passes test
- Electroendosmosis (-Mr):0.15 - 0.19
- Sulphates:Max. 0.20 %
- Residue on ignition:Max. 1.0 %
- Loss on drying:Max. 10.0 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC443023R spec_VWRC443023R spec_VWRC443024S spec_VWRC443025T spec_VWRC443025T
Product specification
More