Agarose
Catalog # A1091.0250
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
Melting Pt:60…90 °C
Storage Temperature:Ambient
EINECS:232-731-8
CAS Number:9012-36-6
MDL Number:MFCD00081294