Agarose D1-LE for molecular biology
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9012-36-6
:
MDL Number:MFCD00081294
Melting Pt:60…90 °C
Storage Temperature:Ambient
EINECS:232-731-8
Related Information
Certificates
Search by Lot NumberALFAH26855.22_TDS_EN
Product specification
Organic Building Blocks and Intermediates