Order Entry
Germany
ContactUsLinkComponent
Agarose for electrophoresis, for electrophoresis medium electroendosmosis (EEO) (Mr = 0.16 - 0.19)
Agarose for electrophoresis, for electrophoresis medium electroendosmosis (EEO) (Mr = 0.16 - 0.19)
Certificates
undefined
Agarose for electrophoresis, for electrophoresis medium electroendosmosis (EEO) (Mr = 0.16 - 0.19)

Specification Test Results

  • Appearance:
    White to off-white to cream Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    86.5 to 89.5 °C (1.5 % gel)
  • Loss on drying:
    ≤10 % (5 g, 110 °C, 24 h)
  • Gel strength:
    ≥750 g/cm² (1 % gel)
  • Gel point:
    34.5 to 37.5 °C (1.5 % gel)
  • Electroendosmosis:
    0.16 to 0.19 (-Mr)
  • Ash:
    ≤0.5 %
  • Sulfate (SO4):
    ≤0.25 %

Specification Test Results

Related Information