Agarose for electrophoresis medium-low electroendosmosis (EEO) (Mr = 0.12)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9012-36-6
:
MDL Number:MFCD00081294
Melting Pt:60 - 90 °C
Storage Temperature:Ambient
EINECS:232-731-8
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:86.5 to 89.5 °C (1.5% gel)
- Water:≤7 % (K.F.)
- Gel strength:≥1200 g/cm² (1% gel)
- Gel point:34.5 to 37.5 °C (1.5% gel)
- Electroendosmosis:0.05 to 0.13 (EEO)
- Ash:≤0.5 %
- Sulfate (SO4):≤0.2 %