Agarose, powder, pure for biochemistry (Mr = - 0.17)
:9012-36-6
:
:MFCD00081294
:60 - 90 °C
:Ambient
:232-731-8
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤7 % (5 g, 110°C, 24 h)
- Gel strength:≥2000 g/cm² (1.5 % gel)
- Gel point:34.5 to 37.5°C (1.5 % gel)
- Electroendosmosis:0.16 to 0.19 (-Mr)
- Ash:≤0.5 %
- Sulfate (SO4):≤0.25 %