Agar
Marketsource
Supplier: Serva
Jump to:
About this item
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- Storage Temperature:Ambient
- EINECS:232-658-1
2 Options Available Below
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- Storage Temperature:Ambient
- EINECS:232-658-1



