Order Entry
Germany
ContactUsLinkComponent
Cellulose
Cellulose
Catalog # SERA14205.02
Supplier:  Serva
CAS Number:  
Cellulose
Catalog # SERA14205.02
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :342.3 g/mol
Merck Index:13,01977
Melting Pt:260…270 °C
Density:1.27…1.6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-674-9
CAS Number:9004-34-6
Formula:(C₁₂H₂₂O₁₁)n
MDL Number:MFCD00081512