Tabletting aid K (cellulose powder), EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, E460ii, SAFC®
Catalog # 1.08197.9020
Supplier: Merck
CAS Number:
Specifications
- Pack type:Cardboard carton
- Pk:20 kg
- Seq:4241
Specifications
About this item
:
:342.3 g/mol
Merck Index:13,01977
Melting Pt:260…270 °C
Density:1.27…1.6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-674-9
CAS Number:9004-34-6
Formula:(C₁₂H₂₂O₁₁)n
MDL Number:MFCD00081512