Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cellulose
Cellulose, microcrystalline
Cellulose, microcrystalline
:
Thermo Fisher Scientific
Cellulose, microcrystalline
:
Thermo Fisher Scientific
:
9004-34-6
:
:
(C₁₂H₂₂O₁₁)n
:
MFCD00081512
:
342.3 g/mol
:
14,1965
:
260…270 °C
:
1.27…1.6 g/cm³ (20 °C)
:
232-674-9
:
GHS07
:
Ambient
msds_ALFAA17730
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Thermo Scientific Cell Culture Workflow