Streptomycin sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3810-74-0
:
Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MDL Number:MFCD00037023
:1457.4 g/mol
Merck Index:12,08983
Melting Pt:250 °C
EINECS:223-286-0
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS08
:GHS07
Storage Temperature:Refrigerator