
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water
Catalog # SERA37241.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
Boiling Pt:∼100 °C (1013 hPa)
Density:∼1.01 g/cm³ (20 °C)
:GHS05
Storage Temperature:Ambient
CAS Number:9002-93-1
:9036-19-5
MDL Number:MFCD00132505