D-(+)-Xylose, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:58-86-6
:
Formula:C₅H₁₀O₅
MDL Number:MFCD00151475
:150.13 g/mol
Merck Index:13,10142
Boiling Pt:331 °C (1013 hPa)
Melting Pt:154 °C
Density:1.525 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-400-7