ContactUsLinkComponent
D-(+)-Xylose
  EHERC17946000
 :  
undefined
D-(+)-Xylose
  EHERC17946000
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029
 :
 :150.13 g/mol
 :13,10142
 :331 °C (1013 hPa)
 :154 °C
 :1.525 g/cm³ (20 °C)
 :Ambient
 :200-400-7
 :58-86-6
 :C₅H₁₀O₅
 :MFCD00151475