Order Entry
Germany
Orders LinkContactUsLinkComponent
 

D-(+)-Mannose, Millipore®

Supplier:   Merck
D-(+)-Mannose, Millipore®
Supplier:  Merck
Certificates
D-(+)-Mannose, Millipore®

2 Options Available Below

Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:133 °C
  • Density:1.539 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 60,40 €
    60,40 €
    1 * 25 g
       60,40 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 212,00 €
    212,00 €
    1 * 100 g
       212,00 €