D-(+)-Glucose, anhydrous 97.5-102.0% Ph. Eur., USP
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Merck Index:14,04459
Melting Pt:146 °C
Density:1.54 g/cm³ (20 °C)
:180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
Specification Test Results
- Appearance::97,5 - 102,0 %
- White or almost white crystalline powder:Passes test
- Heavy metals (as Pb):Max. 5 μg/g
- Residual solvents:Unlikely by manuf.process
- Residues of metal catalysts or reagents:Unlikely by manuf.process
- As (Arsenic):Max. 2 ppm
- Cd (Cadmium):Max. 1 ppm
- Co (Cobalt):Max. 1 ppm
- Hg (Mercury):Max. 2 ppm
- Ni (Nickel):Max. 1 ppm
- Pb (Lead):Max. 2 ppm
- V (Vanadium):Max. 1 ppm
- Conforms to Ph.Eur./USP:Passes test
- Ph.Eur. specifications::Max. 0,5 %
- Assay (on anhydrous) [Ph.Eur.]:97.5 --> 102.0 %
- Identification A [Ph.Eur.]:52.5 --> 53.3 °
- Identification B:Passes test Ph.Eur.
- Identification E:Passes test Ph.Eur.
- Appearance of solution:Passes test Ph.Eur.
- Conductivity [Ph.Eur.]:Max. 20 μS/cm
- Maltose and isomaltose [Ph.Eur.]:Max. 0.4 %
- Maltotriose [Ph.Eur.]:Max. 0.2 %
- Fructose [Ph.Eur.]:Max. 0.15 %
- Unspecified impur. (for each) [Ph.Eur.]:Max. 0.10 %
- Total impurities [Ph.Eur.]:Max. 0.5 %
- Disregard limit [Ph.Eur.]:Max. 0.05 %
- Dextrin:Passes test Ph.Eur.
- Soluble starch, sulfite [Ph.Eur.]:Max. 15 ppm
- Water [Ph.Eur.]:Max. 1.0 %
- USP specifications::Conforms to EMEA/CHMP/SWP/4446/2000
- Assay (calculated on anhydrous) [USP]:97.5 --> 102.0 %
- Identification A:Passes test USP
- Identification C [USP]:Max. 1.0 %
- Maltose and isomaltose [USP]:Max. 0.4 %
- Maltotriose [USP]:Max. 0.2 %
- Fructose [USP]:Max. 0.15 %
- Unspecified impurities (for each) [USP]:Max. 0.10 %
- Total impurities [USP]:Max. 0.5 %
- Disregard limit [USP]:Max. 0.05 %
- Colour and clarity of solution:Passes test USP
- Conductivity (25°C) [USP]:Max. 20 μS/cm
- Soluble starch, sulfites [USP]:Max. 15 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 30L Plastic bucket UN Z (ZPACK13247037) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
284504S_D(+)-GLUCOSE ANHYDROUS PH.EUR.USP_BSE_REV0.pdf 28450_D(+-GLUCOSE ANHYDROUS PH.EUR_USP_BSE_Rev1
BSE/TSE Certificates
28450_D(+-GLUCOSE ANHYDROUS PH.EUR_USP_ALLERGEN_REV0 Allergen_28450_D(+-GLUCOSE ANHYDROUS PH.EUR_USP
ALLERGEN
D-(+)-Glucose, Anhydrous 97,5 - 102% Ph. Eur., USP
Additional Product Details
spec_VWRC284504S spec_VWRC284504S spec_VWRC284508W spec_VWRC284508W spec_VWRC28450BF
Product specification
More