D-(+)-Glucose, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Merck Index:14,04459
Melting Pt:146 °C
Density:1.54 g/cm³ (20 °C)
:180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
Specification Test Results
- IR Spectrum:Passes test
- Specific optical rotation (10 %; water):52.5 - 53.3 °
- Ignition residue (SO₄):Max. 0.03 %
- Insolubility in water:Max. 30 ppm
- Water:Max. 0.5 %
- Cl (Chloride):Max. 25 ppm
- SO₃ (as SO₂):Max. 5 ppm
- SO₄ (Sulphate):Max. 25 ppm
- As (Arsenic):Max. 0.2 ppm
- Cu (Copper):Max. 1 ppm
- Fe (Iron):Max. 1 ppm
- Pb (Lead):Max. 0.5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number101174Y_D(+)-GLUCOSE ANHYDROUS_BSE_REV0.pdf 101175P_ D(+)-GLUCOSE ANHYDROUS ANALAR NORMAPUR_BSE_REV0.pdf 101176K_D(+)-GLUCOSE ANHYDROUS_BSE_REV0.pdf