

Orthophosphoric acid 33% (w/w), Biotech Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Merck Index:13,08966
Melting Pt:169…170 °C
Density:1.29 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-334-9
ADR:8,III
:Filtered through a 0.2 µm filter.
UN:1805
:GHS05
:GHS07
Specification Test Results
- GMP Manufactured Product:32.0 - 34.0%
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- Assay (H₃PO₄):32.0 - 34.0 %
- Alkali Phosphates:Passes Test
- ACS - Heavy Metals (as Pb):≤5 ppm
- Nitrate (NO₃):Passes Test
- Sulfate (SO₄):Passes Test
- Identification:Passes Test
- Phosphorous or Hypophosphorous Acid:Passes Test
- Arsenic (As):≤0.200 ppm
- Appearance of Solution:Passes Test
- Chloride (Cl):≤6 ppm