Order Entry
Germany
ContactUsLinkComponent
D-(+)-Raffinose pentahydrate
D-(+)-Raffinose pentahydrate
Catalog # EHERC16806000
Supplier:  EHRENSTORFER
CAS Number:  
D-(+)-Raffinose pentahydrate
Catalog # EHERC16806000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :594.52 g/mol
Merck Index:12,08279
Melting Pt:80 °C
Density:1.516 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-146-9
CAS Number:17629-30-0
Formula:C₁₈H₃₂O₁₆·5H₂O
MDL Number:MFCD00071590