D-(-)-Sorbitol 70% ≥70% in aqueous solution
Catalog # 1.03576.9270
Supplier: Merck
CAS Number: 50-70-4
Jump to:
Product Details & Documents
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Density:1.3 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-061-5
Specifications
Specifications
- Pk:270 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



