D-(-)-Sorbitol F, liquid E420, SAFC®
Catalog # 1.02993.9903
Supplier: Merck
CAS Number:
Specifications
- Pk:1.000 kg
- Seq:4876
Specifications
About this item
:
Density:1.3 g/cm³ (20 °C)
:182.17 g/mol
Storage Temperature:Ambient
EINECS:200-061-5
CAS Number:50-70-4
:98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708