Order Entry
Germany
ContactUsLinkComponent
D-(-)-Sorbitol ≥70% in aqueous solution
D-(-)-Sorbitol ≥70% in aqueous solution
Catalog # KRAF10095.4700
Supplier:  Bernd Kraft
CAS Number:  
undefined
D-(-)-Sorbitol ≥70% in aqueous solution
Catalog # KRAF10095.4700
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL
  • Seq:
    2895

Specifications

About this item

 :
Density:1.3 g/cm³ (20 °C)
 :182.17 g/mol
Storage Temperature:Ambient
EINECS:200-061-5
CAS Number:50-70-4
 :98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708