
L(+)-Lactic acid ≥85% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-33-4
:
Formula:C₃H₆O₃
MDL Number:MFCD00064266
:90.08 g/mol
Density:1.21 g/cm³ (20 °C)
EINECS:201-196-2
:GHS05
Storage Temperature:Refrigerator
Frequently Bought Together