cis-Platin, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:15663-27-1
:
Formula:H₆Cl₂N₂Pt
MDL Number:MFCD00011623
:300.05 g/mol
Merck Index:13,02341
Density:3.738 g/cm³ (20 °C)
EINECS:239-733-8
UN:3288
ADR:6.1,II
:M-Clarity™ quality level = MQ200