Order Entry
Germany
Orders LinkContactUsLinkComponent
Boron trifluoride diethyl ether complex

Boron trifluoride diethyl ether complex

Glass bottle, 1 L
Catalog # MOLEM20561377
Flammable
Toxic
Corrosive
Health
Marketsource
Supplier:   Molekula
Boron trifluoride diethyl ether complex
Catalog # MOLEM20561377
Supplier:  Molekula
Marketsource
CAS Number:  109-63-7
Boron trifluoride diethyl ether complex

Glass bottle, 1 L

CAS Number:  109-63-7
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
  • CAS Number:109-63-7
  • Formula:C₄H₁₀BF₃O
  • MDL Number:MFCD00013194
  • Molecular Weight:141.93 g/mol
  • Merck Index:13,01340
  • Boiling Pt:126 °C (1013 hPa)
  • Melting Pt:–60 °C
  • Density:1.125 g/cm³ (25 °C)
  • Flash Pt:59 °C
  • Storage Temperature:Ambient
  • EINECS:203-689-8
  • UN:2604
  • ADR:8,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.